This product is now discontinued, View Related Products
WorkBeads™ Size Exclusion Purification Media
Separate molecules based on their size by filtration through the gel
  • Great selectivity and efficiency results in excellent resolution
  • High chemical stability for easy clean-in-place (CIP)
Separation by size exclusion occurs when different size molecules are included or excluded from the pores of the matrix. Larger molecules, too big to enter the pores, flow quickly down the column while smaller-sized molecules enter the pores and pass through, resulting in slower progress through the media. This method is great for separating large molecules from the small molecules while preserving the biological activity of the particles to be separated. Recommended flow rate of 600 cm/hr. Stable over pH 2 to 13 working range.
$319.73 - $4,985.95CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cutoff (kDa) Media Volume (mL)
Availability Pricing
Bio-Works WorkBeads 40 SEC Size Exclusion Medium, 50 to 1200 kDa; 25 mL RK-11946-93 Mfr # KN40300001
COMPARE
1200 25
$340.35CAD / Each
Bio-Works WorkBeads 40 SEC Size Exclusion Medium, 50 to 1200 kDa; 300 mL RK-11946-94 Mfr # KN40300003
COMPARE
1200 300
$1,978.76CAD / Each
Bio-Works WorkBeads 40 SEC Size Exclusion Medium, 50 to 1200 kDa; 1 L RK-11946-95 Mfr # KN40300010
COMPARE
1200 1000
$3,942.79CAD / Each
Bio-Works WorkBeads 40/100 SEC Size Exclusion Medium, 10 to 150 kDa; 25 mL RK-11946-96 Mfr # KN40340001
COMPARE
150 25
$330.04CAD / Each
Bio-Works WorkBeads 40/100 SEC Size Exclusion Medium, 10 to 150 kDa; 300 mL RK-11946-97 Mfr # KN40340003
COMPARE
150 300
$1,966.98CAD / Each
Bio-Works WorkBeads 40/100 SEC Size Exclusion Medium, 10 to 150 kDa; 1 L RK-11946-98 Mfr # KN40340010
COMPARE
150 1000
$4,985.95CAD / Each
Bio-Works WorkBeads 40/10 000 SEC Size Exclusion Medium, up to 10,000 kDa; 25 mL RK-11946-99 Mfr # KN40350001
COMPARE
10000 25
$319.73CAD / Each
Bio-Works WorkBeads 40/10 000 SEC Size Exclusion Medium, up to 10,000 kDa; 300 mL RK-11947-00 Mfr # KN40350003
COMPARE
10000 300
$1,986.13CAD / Each
Bio-Works WorkBeads 40/10 000 SEC Size Exclusion Medium, up to 10,000 kDa; 1 L RK-11947-01 Mfr # KN40350010
COMPARE
10000 1000
$4,985.95CAD / Each
Showing 1- 9 of 9