This product is now discontinued, View Related Products
WorkBeads™ Ion Exhange Purification Media
High-throughput separation of proteins, peptides, and oligonucleotides
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • High chemical stability for easy clean-in-place (CIP)
  • Efficient separation due to the narrow particle size distribution
Manufactured using agarose-based chromatographic resins, WorkBeads ion exchange purification media provide a tight size distribution and exceptional mechanical stability. Average bead size of 40 µm enables high-resolution separation at flows up to 700 cm/hr at moderate backpressure. Stable over pH 2 to 13 range.
$350.67 - $4,644.13CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials Media Volume (mL)
Availability Pricing
Bio-Works WorkBeads 40 Q Ion Exchange Medium, Quaternary Amine; 25 mL RK-11946-84 Mfr # KN40100001
COMPARE
Quaternary amine resin 25
$352.14CAD / Each
Bio-Works WorkBeads 40 Q Ion Exchange Medium, Quaternary Amine; 200 mL RK-11946-85 Mfr # KN40100002
COMPARE
Quaternary amine resin 200
$1,121.25CAD / Each
Bio-Works WorkBeads 40 Q Ion Exchange Medium, Quaternary Amine; 1 L RK-11946-86 Mfr # KN40100010
COMPARE
Quaternary amine resin 1000
$4,644.13CAD / Each
Bio-Works WorkBeads 40 DEAE Ion Exchange Medium, Diethylaminoethyl; 25 mL RK-11946-87 Mfr # KN40150001
COMPARE
Diethylaminoethyl resin 25
$350.67CAD / Each
Bio-Works WorkBeads 40 DEAE Ion Exchange Medium, Diethylaminoethyl; 200 mL RK-11946-88 Mfr # KN40150002
COMPARE
Diethylaminoethyl resin 200
$1,150.72CAD / Each
Bio-Works WorkBeads 40 DEAE Ion Exchange Medium, Diethylaminoethyl; 1 L RK-11946-89 Mfr # KN40150010
COMPARE
Diethylaminoethyl resin 1000
$4,094.55CAD / Each
Bio-Works WorkBeads 40 S Ion Exchange Medium, Sulfonate; 25 mL RK-11946-90 Mfr # KN40200001
COMPARE
Sulfonate resin 25
$352.14CAD / Each
Bio-Works WorkBeads 40 S Ion Exchange Medium, Sulfonate; 200 mL RK-11946-91 Mfr # KN40200002
COMPARE
Sulfonate resin 200
$1,121.25CAD / Each
Bio-Works WorkBeads 40 S Ion Exchange Medium, Sulfonate; 1 L RK-11946-92 Mfr # KN40200010
COMPARE
Sulfonate resin 1000
$3,913.32CAD / Each
Showing 1- 9 of 9