WIKA Stainless Steel Digital Gauges
Durable process gauges offer precise pressure readings
  • Accuracy of ±0.25% best-fit-straight-line
Stainless steel digital pressure gauges feature a large seven-segment, four-digit LCD with bar graph for easy and quick pressure readings. Gauges have min/max readings and choice of units: psi, bar, or mPa. Pressure sensor is manufactured to provide high accuracy and long-term stability. Gauges up to 600 psi use ceramic sensor technology. Gauges 1450 psi and higher use thin film sensor technology. Optional rubber boot 68052-90 offers protection against rough use.
$724.19 - $790.02CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 145 psi, 1/4" NPT(M) RK-68052-60 Mfr # 50365495
COMPARE
±0.25% BFSL 145 0.1 psi
$724.19CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 600 psi, 1/4" NPT(M) RK-68052-64 Mfr # 50365517
COMPARE
±0.25% BFSL 600 0.1 psi
$724.19CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 1450 psi, 1/4" NPT(M) RK-68052-66 Mfr # 50365525
COMPARE
±0.25% BFSL 1450 0.1 psi
$790.02CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 3000 psi, 1/4" NPT(M) RK-68052-68 Mfr # 50365592
COMPARE
±0.25% BFSL 3000 1 psi
$790.02CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 5000 psi, 1/4" NPT(M) RK-68052-70 Mfr # 50365614
COMPARE
±0.25% BFSL 5000 1 psi
$790.02CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 7500 psi, 1/4" NPT(M) RK-68052-72 Mfr # 50365622
COMPARE
±0.25% BFSL 7500 1 psi
$790.02CAD / Each
Wika DG-10-S Stainless Steel Digital Gauge ~ 0 to 10,000 psi, 1/4" NPT(M) RK-68052-74 Mfr # 50365631
COMPARE
±0.25% BFSL 10000 1 psi
$790.02CAD / Each
Showing 1- 7 of 7