This product is now discontinued, View Related Products
Wide-Mouth PET Jars
Maintain FDA-compliance
  • Unbreakable design
  • Resist moisture and oxygen to keep samples fresh
These polyethylene terephthalate (PET) jars are highly resistant to impact and scuffing—ideal for your storing needs. Jars feature wide mouths for easy filling, pouring, and cleaning. Unbreakable, jars come with polyethylene linerless caps.
Use these plastic jars instead of glass jars—they weigh 90% less, have equal clarity, and offer excellent barriers to moisture and oxygen. FDA-compliant for food products. Please note, the jars are not leakproof or autoclavable.
$83.25 - $148.08CAD / Pkg of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Outside Diameter (OD) (in) Overall Height (in)
Availability Pricing
COMPARE
180 2 1/4 3 1/2
$90.17CAD / Pkg of 12
COMPARE
240 2 1/4 4 1/2
$83.25CAD / Pkg of 12
COMPARE
350 2 1/2 5
$88.11CAD / Pkg of 12
COMPARE
480 2 3/4 5 7/8
$92.82CAD / Pkg of 12
COMPARE
1000 3 1/2 6
$148.08CAD / Pkg of 12
COMPARE
2000 5 7
$107.56CAD / Pkg of 6
COMPARE
4000 6 1/8 9 3/4
$126.12CAD / Pkg of 4
Showing 1- 7 of 7