WHEATON® V-Vial, Clear Glass with PTFE-Lined Phenolic Caps
Ideal for small scale reactions, storage and shipping
  • Constructed of clear borosilicate glass for easy sample viewing
Manufactured from low extractable borosilicate glass to safeguard against changes in product pH caused by alkali, which can occur with ordinary glass. Sturdy vials feature a conical interior bottom that provides downward drainage for maximum retreival of contents. Includes black phenolic screw caps with PTFE faced white rubber liner.
$275.52 - $363.93CAD / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Tube Color Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 986254 V-Vials, Clear Glass w/PTFE-Lined Penolic Caps, 1.0 mL; 12/Cs RK-08925-74 Mfr # W986254NG
COMPARE
1 47/64 Clear 1
$275.52CAD / Case of 12
DWK Life Sciences (Wheaton) W986256NG V-Vials, Clear Glass w/PTFE-Lined Penolic Caps, 2.0 mL; 12/Cs RK-08925-76 Mfr # W986256NG
COMPARE
2 2/5 Clear 2
$331.51CAD / Case of 12
DWK Life Sciences (Wheaton) 986259 V-Vials, Clear Glass w/PTFE-Lined Penolic Caps, 5.0 mL; 12/Cs RK-08925-82 Mfr # W986259NG
COMPARE
2 9/16 Clear 5
$363.93CAD / Case of 12