This product is now discontinued, View Related Products
WHEATON Cryule Plastic Cryogenic Vials
Leakproof even during changing temperatures—ultimate protection during transport
  • Made from low binding, cryogenic-grade virgin polypropylene
  • Lot-certified vials are free of RNase/DNase and Endotoxin—ensure product integrity
Tubes and closures have same coefficient of expansion to enhance leakproof qualities. White marking area aids in easy sample identification. Vials withstand temperatures from 121°C to 196°C (250 to 384°F). Flat-bottom vials have conical shaped wells and are self standing. Exacting round-bottom vials allow centrifugation up to 17,000 max RCF (xG). Order racks separately at left. Autoclavable.
$340.35 - $642.40CAD / Case
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Cap color Tube Bottom Shape Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 985721 Cryogenic Vial, 2mL, Round Bottom RK-03756-01 Mfr # 985721
COMPARE
White Round 2
$642.40CAD / Case
DWK Life Sciences (Wheaton) 985726 Cryogenic Vial, non-Sterile 2mL, Round Bottom without white marking area RK-03756-02 Mfr # 985726
COMPARE
White Round 2
$545.89CAD / Case
DWK Life Sciences (Wheaton) W985860 Cryogenic Vial, Sterile 1 mL, Round Bottom RK-03756-04 Mfr # W985860
COMPARE
Natural Round 1.2
$421.39CAD / Case of 500
DWK Life Sciences (Wheaton) 985735 Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Red Cap RK-03756-11 Mfr # 985735
COMPARE
Red Flat 2
$340.35CAD / Case
DWK Life Sciences (Wheaton) 985736 Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Pink Cap RK-03756-12 Mfr # 985736
COMPARE
Pink Flat 2
$340.35CAD / Case
DWK Life Sciences (Wheaton) W985686 Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Blue Cap RK-03756-18 Mfr # W985868
COMPARE
Blue Flat 2
$403.71CAD / Case of 500