CYTIVA Whatman™ Quantitative Filter Papers
The industry standard for high-purity filtration
  • Available in a variety of grades and sizes
Our CYTIVA Whatman™ filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box.
Quantitative filter papers are the choice for gravimetric analysis. High-purity hardened filters have a high wet-strength and are chemical-resistant to handle vacuum filtration or acid/alkali solutions.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$44.35 - $429.12CAD / Box of 100
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material Pore Size (micron)
Availability Pricing
Whatman 1441-042 41 Ashless Quantitative Filter Paper, 20-25um, 4.25cm; 100/Box RK-06647-00 Mfr # 1441-042
COMPARE
4.25 Cotton Fiber 23
$44.35CAD / Box of 100
Whatman 1441-055 41 Ashless Quantitative Filter Paper, 20-25um, 5.5cm; 100/Box RK-06647-01 Mfr # 1441-055
COMPARE
5.5 Cotton Fiber 23
$47.15CAD / Box of 100
Whatman 1441-090 41 Ashless Quantitative Filter Paper, 20-25um, 9cm; 100/Box RK-06647-02 Mfr # 1441-090
COMPARE
9 Cotton Fiber 23
$81.04CAD / Box of 100
Whatman 1441-110 41 Ashless Quantitative Filter Paper, 20-25um, 11cm; 100/Box RK-06647-03 Mfr # 1441-110
COMPARE
11 Cotton Fiber 23
$109.91CAD / Box of 100
Whatman 1441-125 41 Ashless Quantitative Filter Paper, 20-25um, 12.5cm; 100/Box RK-06647-04 Mfr # 1441-125
COMPARE
12.5 Cotton Fiber 23
$125.09CAD / Box of 100
Whatman 1441-150 41 Ashless Quantitative Filter Paper, 20-25um, 15cm; 100/Box RK-06647-05 Mfr # 1441-150
COMPARE
15 Cotton Fiber 23
$151.76CAD / Box of 100
Whatman 1441-185 41 Ashless Quantitative Filter Paper, 20-25um, 18.5cm; 100/Box RK-06647-06 Mfr # 1441-185
COMPARE
18.5 Cotton Fiber 23
$248.27CAD / Box of 100
Whatman 1441-240 41 Ashless Quantitative Filter Paper, 20-25um, 24cm; 100/Box RK-06647-07 Mfr # 1441-240
COMPARE
24 Cotton Fiber 23
$370.19CAD / Box of 100
Whatman 1440-042 40 Ashless Quantitative Filter Paper, 8um, 4.25cm; 100/Box RK-06647-10 Mfr # 1440-042
COMPARE
4.25 Cotton Fiber 8
$52.60CAD / Box of 100
Whatman 1440-055 40 Ashless Quantitative Filter Paper, 8um, 5.5cm; 100/Box RK-06647-11 Mfr # 1440-055
COMPARE
5.5 Cotton Fiber 8
$58.64CAD / Box of 100
Whatman 1440-090 40 Ashless Quantitative Filter Paper, 8um, 9cm; 100/Box RK-06647-12 Mfr # 1440-090
COMPARE
9 Cotton Fiber 8
$85.60CAD / Box of 100
Whatman 1440-110 40 Ashless Quantitative Filter Paper, 8um, 11cm; 100/Box RK-06647-13 Mfr # 1440-110
COMPARE
11 Cotton Fiber 8
$101.37CAD / Box of 100
Whatman 1440-125 40 Ashless Quantitative Filter Paper, 8um, 12.5cm; 100/Box RK-06647-14 Mfr # 1440-125
COMPARE
12.5 Cotton Fiber 8
$120.67CAD / Box of 100
Whatman 1440-150 40 Ashless Quantitative Filter Paper, 8um, 15cm; 100/Box RK-06647-15 Mfr # 1440-150
COMPARE
15 Cotton Fiber 8
$167.23CAD / Box of 100
Whatman 1440-185 40 Ashless Quantitative Filter Paper, 8um, 18.5cm; 100/Box RK-06647-16 Mfr # 1440-185
COMPARE
18.5 Cotton Fiber 8
$219.17CAD / Box of 100
Whatman 1440-240 40 Ashless Quantitative Filter Paper, 8um, 24cm; 100/Box RK-06647-17 Mfr # 1440-240
COMPARE
24 Cotton Fiber 8
$381.61CAD / Box of 100
Whatman 1442-042 42 Ashless Quantitative Filter Paper, 2.5um, 4.25cm; 100/Box RK-06647-20 Mfr # 1442-042
COMPARE
4.25 Cotton Fiber 2.5
$47.15CAD / Box of 100
Whatman 1442-055 42 Ashless Quantitative Filter Paper, 2.5um, 5.5cm; 100/Box RK-06647-21 Mfr # 1442-055
COMPARE
5.5 Cotton Fiber 2.5
$59.08CAD / Box of 100
Whatman 1442-090 42 Ashless Quantitative Filter Paper, 2.5um, 9cm; 100/Box RK-06647-22 Mfr # 1442-090
COMPARE
9 Cotton Fiber 2.5
$97.54CAD / Box of 100
Whatman 1541-090 541 Ashless Quantitative Filter Paper, 20-25um, 9cm; 100/Box RK-06647-32 Mfr # 1541-090
COMPARE
9 Cellulose 23
$97.54CAD / Box of 100
Showing 1 - 20 of 42