CYTIVA Whatman™ Qualitative Filter Papers
The industry standard for high-purity filtration
  • Available in a variety of grades and sizes
Our Whatman® filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box.
Qualitative filter papers are used for liquid clarifications, analytical separations, or air and water analysis.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$20.19 - $210.33CAD / Box of 100
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Pore Size (micron)
Availability Pricing
Whatman 1004-042 Qualitative Filter Papers; 4.25 cm dia; pore size, 20-25µ; 100/box RK-06648-00 Mfr # 1004-042
COMPARE
4.25 23
$20.19CAD / Box of 100
Whatman 1004-055 Qualitative Filter Papers; 5.5 cm dia; pore size, 20-25µ; 100/box RK-06648-01 Mfr # 1004-055
COMPARE
5.5 23
$25.78CAD / Box of 100
Whatman 1004-090 Qualitative Filter Papers; 9.0 cm dia; pore size, 20-25 µ; 100/box RK-06648-02 Mfr # 1004-090
COMPARE
9 23
$29.17CAD / Box of 100
Whatman 1004-110 Qualitative Filter Papers; 11.0 cm dia; pore size, 20-25 µ; 100/box RK-06648-03 Mfr # 1004-110
COMPARE
11 23
$37.42CAD / Box of 100
Whatman 1004-125 Qualitative Filter Papers; 12.5 cm dia; pore size, 20-25 µ; 100/box RK-06648-04 Mfr # 1004-125
COMPARE
12.5 23
$50.10CAD / Box of 100
Whatman 1004-150 Qualitative Filter Papers; 15 cm dia; pore size, 20-25 µ; 100/box RK-06648-05 Mfr # 1004-150
COMPARE
15 23
$60.56CAD / Box of 100
Whatman 1004-185 Qualitative Filter Papers; 18.5 cm dia; pore size, 20-25 µ; 100/box RK-06648-06 Mfr # 1004-185
COMPARE
18.5 23
$98.86CAD / Box of 100
Whatman 1004-240 Qualitative Filter Papers; 24 cm dia; pore size, 20-25 µ; 100/box RK-06648-07 Mfr # 1004-240
COMPARE
24 23
$180.12CAD / Box of 100
Whatman 1001-042 Qualitative Filter Papers; 4.25 cm dia; pore size, 11µ; 100/box RK-06648-10 Mfr # 1001-042
COMPARE
4.25 11
$23.43CAD / Box of 100
Whatman 1001-055 Qualitative Filter Papers; 5.5 cm dia; pore size, 11µ; 100/box RK-06648-11 Mfr # 1001-055
COMPARE
5.5 11
$22.54CAD / Box of 100
Whatman 1001-090 Qualitative Filter Papers; 9.0 cm dia; pore size, 11 µ; 100/box RK-06648-12 Mfr # 1001-090
COMPARE
9 11
$28.29CAD / Box of 100
Whatman 1001-110 Qualitative Filter Papers; 11.0 cm dia; pore size, 11 µ; 100/box RK-06648-13 Mfr # 1001-110
COMPARE
11 11
$34.18CAD / Box of 100
Whatman 1001-125 Qualitative Filter Papers; 12.5 cm dia; pore size, 11 µ; 100/box RK-06648-14 Mfr # 1001-125
COMPARE
12.5 11
$35.51CAD / Box of 100
Whatman 1001-150 Qualitative Filter Papers; 15 cm dia; pore size, 11 µ; 100/box RK-06648-15 Mfr # 1001-150
COMPARE
15 11
$50.10CAD / Box of 100
Whatman 1001-185 Qualitative Filter Papers; 18.5 cm dia; pore size, 11 µ; 100/box RK-06648-16 Mfr # 1001-185
COMPARE
18.5 11
$71.46CAD / Box of 100
Whatman 1001-240 Qualitative Filter Papers; 24 cm dia; pore size, 11 µ; 100/box RK-06648-17 Mfr # 1001-240
COMPARE
24 11
$116.55CAD / Box of 100
Whatman 1005-042 Qualitative Filter Papers; 4.25 cm dia; pore size, 2.5 µ; 100/box RK-06648-20 Mfr # 1005-042
COMPARE
4.25 2.5
$22.40CAD / Box of 100
Whatman 1005-055 Qualitative Filter Papers; 5.5 cm dia; pore size, 2.5 µ; 100/box RK-06648-21 Mfr # 1005-055
COMPARE
5.5 2.5
$24.02CAD / Box of 100
Whatman 1005-090 Qualitative Filter Papers; 9.0 cm dia; pore size, 2.5 µ; 100/box RK-06648-22 Mfr # 1005-090
COMPARE
9 2.5
$38.16CAD / Box of 100
Whatman 1005-110 Qualitative Filter Papers; 11.0 cm dia; pore size, 2.5 µ; 100/box RK-06648-23 Mfr # 1005-110
COMPARE
11 2.5
$54.22CAD / Box of 100
Showing 1 - 20 of 24