CYTIVA Whatman™ Disposable Capsule Filters, Polypropylene Membrane
Use polypropylene (PP) membrane filters for solvents, clean air/gas, prefiltration, and sample preparation
  • Autoclavable
Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$124.65 - $191.54CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman 6703-3610 Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk RK-29700-20 Mfr # 6703-3610
COMPARE
Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8\" ID; 1/Pk
$132.61CAD / Each
Whatman 6703-3650 Polycap HD PP Capsule Filter, 5.0um, 385cm2, 1/4-3/8" ID; 1/Pk RK-29700-22 Mfr # 6703-3650
COMPARE
Polycap HD PP Capsule Filter, 5.0um, 385cm2, 1/4-3/8\" ID; 1/Pk
$134.96CAD / Each
Whatman 6703-7510 Polycap HD PP Capsule Filter, 1.0um, 805cm2, 1/2" ID; 1/Pk RK-29700-30 Mfr # 6703-7510
COMPARE
Polycap HD PP Capsule Filter, 1.0um, 805cm2, 1/2\" ID; 1/Pk
$191.54CAD / Each
Whatman 6703-7550 Polycap HD PP Capsule Filter, 5.0um, 805cm2, 1/2" ID; 1/Pk RK-29700-32 Mfr # 6703-7550
COMPARE
Polycap HD PP Capsule Filter, 5.0um, 805cm2, 1/2\" ID; 1/Pk
$191.54CAD / Each
Whatman 6703-3611 Disposable Capsule Filters, PP membrane, 10um, 1/4"-3/8" stepped barb RK-29701-40 Mfr # 6703-3611
COMPARE
Disposable Capsule Filters, PP membrane, 10um, 1/4\"-3/8\" stepped barb
$124.65CAD / Pkg of 1
Whatman 6703-7511 Disposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb RK-29701-42 Mfr # 6703-7511
COMPARE
Disposable Capsule Filters, PP membrane, 10um, 1/2\" stepped barb
$146.16CAD / Pkg of 1