Waterless Ice Bucket Kit
Avoid contamination risk – dry beads provide ice-free cooling
  • Several hours of constant cooling
  • Durable and lightweight — polyurethane bucket with molded side knobs
Keep your samples cold, clean, and dry. This ice-free bucket helps you avoid water puddles and float-away tubes. Use anywhere in place of traditional ice. Simply cool the beads and chill packs in your refrigerator or freezer, then assemble the chill bucket. Put the chill packs on the bottom, and place filled bead bag in the bucket. Create your own temperature range by starting with beads at temperatures at ambient, or cooled to 4°C, -20°C, or -80°C. May substitute chill packs with dry ice.
$1,035.06CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterless ice bucket with beads; 4L capacity
$1,035.06CAD / Each