Vortex-Genie Pulse Shakers
True pulsing action providing more collisions and random motion
  • Ideal for applications requiring more aggressive action than standard vortexing.
  • Fully programmable the variable run/stop time provides quick pulsing or interval mixing to keep samples in suspension, or use in continuous mode for vortexing.
  • Featuring digital control and display of speed and time, this shaker will deliver reproducible and accurate mixing.
  • Accepts all standard Genie attachments (sold separately).
$1,135.98CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-P236 Pulse Shaker, 500 to 3000 rpm, 120 VAC RK-04724-38 Mfr # SI-P236
COMPARE
120
$1,135.98CAD / Each
Scientific Industries Genie SI-P246 Pulse, 230 VAC, Without Plug. RK-04724-39 Mfr # SI-P246
COMPARE
230
$1,135.98CAD / Each