This product is now discontinued, View Related Products
Vee Gee Battery/Coolant Hand Refractometers
  • Anti-freeze coolant testing/battery recharge level
  • Largest display available
  • Durable, all-metal design
It's essential to know the freezing point of anti-freeze coolants for vehicular safety. Keeping a suitable concentration of coolant prevents engine trouble and possible radiator corrosion. These refractometers (available with either °F or °C ranges) monitor the freezing points of both ethylene glycol and propylene glycol; in addition, they indicate battery recharge level.
Simple to use and virtually an indispensable instrument for gas stations, service shops, cab companies, etc., both refractometer models feature automatic temperature compensation (ATC); an ideal choice for testing in conditions with large ambient temperature fluctuations.
$197.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Brix Range (%) Resolution
Availability Pricing
Vee Gee 43062 Coolant/Battery Acid Refractometer; -50 to 0C, 1.10 to 1.40 SG RK-26048-07 Mfr # 43062
COMPARE
0.01 (nD)
$197.43CAD / Each
Vee Gee 43062 Coolant/Battery Acid Refractometer; -60 to 32F, 1.10 to 1.40 SG RK-26048-59 Mfr # 43064
COMPARE
0.01 (nD)
$197.43CAD / Each