Vaisala Intrinsically Safe Humidity and Temperature Transmitters
Factory Mutual approved for hazardous indoor applications
  • Meets Class I, II, III, Division 1, Groups A-G and Division 2, Groups A-D, F and G
Measure temperature and humidity in environmentally hazardous areas. NEMA 4 (IP65) housing is rated water- and dust-proof. Wall-mount and duct-mount versions are available with or without an LCD. Two 4 to 20 mA outputs offer analog signals, one for humidity and one for temperature.
What’s included: certificate of NIST traceability. Wall-mount transmitters include a probe measuring 4 5/8”L x 1/2” dia. Duct-mount transmitters include a probe measuring 2 11/16”L x 1/2” dia with 78" (2 m) of cable length, and a duct installation kit.
$6,567.18CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RH Range (%) Max Temperature (° F) Mounting Type
Availability Pricing
Vaisala HMT370 Wall-Mount Humidity Transmitter With Display RK-37510-12 Mfr # HMT370EX 15A1NB2AC1AN
COMPARE
100 140 Wall
$6,567.18CAD / Each