This product is now discontinued, View Related Products
Vaisala Combined Pressure/Humidity/Temperature Transmitter
Barometric pressure, humidity and temperature measurement in one transmitter
  • Vaisala sensors provide proven, accurate, and reliable measurement with excellent long-term stability
  • Three analog outputs and a serial interface
  • NIST-traceable calibration
The Vaisala PTU-series transmitters are designed for demanding industrial and laboratory applications where multiple environmental conditions need to be monitored. Typical applications include environmental monitoring in calibration laboratories, industrial test cells, and weather stations. The PTU transmitter uses Vaisala's BAROCAP® sensor for pressure, HUMICAP® sensor for humidity, and a platinum RTD sensor for temperature-all known for their high accuracy and excellent long-term stability.
These versatile, multipurpose transmitters feature three analog outputs and a RS-232 serial interface which is compatible with major GPS receivers and NMEA-coded messages. Models 37463-12 and-14 feature a graphic/numeric display and an easy-to-use, menu-based user interface. All models having da
$4,823.88 - $5,531.84CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Display Type Max RH Range (%) Max Temperature (° F)
Availability Pricing
Vaisala PTU300 Pressure, RH/, and Temperature Transmitter (No Display - Wall Mount) RK-37463-11 Mfr # PTU30012A00A1BCPB1B0A1AAD0B0A
COMPARE
None 100 140
$4,823.88CAD / Each
Vaisala PTU300 Pressure, RH/, and Temperature Transmitter (Display - Wall Mount) RK-37463-12 Mfr # PTU30012A01A1BCPB1B0A1AAD0B0A
COMPARE
LCD 100 140
$5,096.46CAD / Each
Vaisala PTU300 Pressure, RH/, and Temperature Transmitter (No Display - Remote Probe) RK-37463-13 Mfr # PTU30032B00A1BCPB1B0A1AAB0B0A
COMPARE
None 100 140
$5,076.57CAD / Each
Vaisala PTU300 Pressure, RH/, and Temperature Transmitter (Display - Remote Probe) RK-37463-14 Mfr # PTU30032B01A1BCPB1B0A1AAB0B0A
COMPARE
LCD 100 140
$5,531.84CAD / Each