Cole-Parmer® Vacuum/Pressure Pumps Coated with PTFE
Wetted parts are coated with PTFE to handle your toughest applications
  • Lightweight aluminum construction and compact size
  • Pumps with gauges and regulators give you more control of your vacuum/pressure applications
These pumps are ideal for benchtop applications such as vacuum filtration. Pumps include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
$802.26 - $1,301.00CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Description
Availability Pricing
COMPARE
0.75 23.2 PTFE-Coated Vacuum Pump, Gauge/Reg/Valve; 0.75 cfm/23.2\"Hg-25psi/115V
$1,256.06CAD / Each
COMPARE
0.55 22 PTFE-Coated Vacuum Pump, Gauge/Reg/Valve; 0.55 cfm/22\"Hg-20psi/220V
$1,301.00CAD / Each
COMPARE
1.2 22.9 PTFE-Coated Vacuum Pump, Gauge/Reg/Valve; 1.2 cfm/22.9\"Hg-20psi/12VDC
$1,295.11CAD / Each
COMPARE
0.75 23.2 Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 115 VAC
$802.26CAD / Each
COMPARE
0.55 22 Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.55 cfm, 220 VAC
$802.26CAD / Each
COMPARE
1.2 22.9 Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 1.2 cfm, 12 VDC
$810.36CAD / Each