UVP White Light Transilluminators
Uniform white light diffusion for viewing negative and film positives
  • Cool white light through an evenly diffused glass surface which permanently sealed into the unit for durability
  • Scratch and chemical resistant plexiglass surface can be cleaned easily
  • Excellent for viewing coomassie blue stained gels as well as autoradiographs, x-rays, and microtiter plates
Two sizes available ¨C 8 x 10¡± film sizes contains two 8-watt tubes, or 14 x 17¡± film sizes contains four 14-watt tubes.
$1,937.51 - $2,593.17CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Filter Length (cm) Filter Width (cm) Power (VAC)
Availability Pricing
UVP 95-0208-01 White Light Transilluminator, 8-Watt, 21 x 26 cm; 115 VAC RK-09814-00 Mfr # 95-0208-01
COMPARE
53.34 66 115
$1,937.51CAD / Each
UVP 95-0214-01 White Light Transilluminator, 14-Watt, 36 x 43 cm; 115 VAC RK-09814-10 Mfr # 95-0214-01
COMPARE
36 43 115
$2,593.17CAD / Each
UVP 95-0214-02 White Light Transilluminator, 14-Watt, 36 x 43 cm; 230 VAC RK-09814-15 Mfr # 95-0214-02
COMPARE
36 43 230
$2,401.63CAD / Each