This product is now discontinued, View Related Products
uVISC Portable Viscometer Viscosity Chips
Only requires 400 microliter sample size!
The microfluidic sensors measure viscosity by the drop in pressure that occurs while the sample flows through a small rectangular flow channel.
$2,212.30 - $2,244.71CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Viscosity Range (cP) Min Viscosity Range (cP)
Availability Pricing
RheoSense HA01-01 Viscosity Chip, 0 to 100 cP Flow channel depth: 50 microns RK-98946-42 Mfr # HA01-01
COMPARE
100 0
$2,244.71CAD / Each
RheoSense HA02-01 Viscosity Chip, 4 to 600 cP Flow channel depth: 100 microns RK-98946-43 Mfr # HA02-01
COMPARE
600 4
$2,223.35CAD / Each
RheoSense HA03-01 Viscosity Chip, 10 to 600 cP Flow channel depth: 200 microns RK-98946-44 Mfr # HA03-01
COMPARE
600 10
$2,233.66CAD / Each
RheoSense HA04-01 Viscosity Chip, 30 to 8000 cP Flow channel depth: 300 microns RK-98946-45 Mfr # HA04-01
COMPARE
8000 30
$2,223.35CAD / Each
RheoSense HB02-01 Viscosity Chip, 60 to 5000 cP Flow channel depth: 100 microns RK-98946-46 Mfr # HB02-01
COMPARE
5000 60
$2,233.66CAD / Each
RheoSense HB03-01 Viscosity Chip, 70 to 6000 cP Flow channel depth: 200 microns RK-98946-47 Mfr # HB03-01
COMPARE
6000 70
$2,244.71CAD / Each
RheoSense HB04-01 Viscosity Chip, 100 to 20,000 cP RK-98946-48 Mfr # HB04-01
COMPARE
20000 100
$2,223.35CAD / Each
RheoSense HC02-01 Viscosity Chip, 400 to 20,000 cP RK-98946-49 Mfr # HC02-01
COMPARE
20000 400
$2,212.30CAD / Each
Showing 1- 8 of 8