Electro-Tech Systems Ultrasonic Humidifier, High-Capacity System
Maintain up to 100% RH at room temperature
  • Fine 5-micron droplets reduce moisture deposition on sensitive items
  • Built-in support for open loop or closed loop operation
  • Universal voltage supply 85 to 305 VAC can be use anywhere
The ultrasonic humidifier is ideal for lab research and industrial processing in medical, pharmaceutical, university, cosmetics and hair, curing, sample conditioning, and other applications. It is designed to minimizes noise and vibration that could impact sensitive measurements.
The system can be utilized in an open-loop or closed-loop configuration and functions with any Electro-Tech Systems controller and compatible chamber or glove box. The system can also be utilized in an open-air environment.
$2,934.99CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
Electro-Tech Systems Ultrasonic Humidifier, High-Capacity System; 85 to 305 VAC RK-37700-66 Mfr # 5482-220V
COMPARE
0.67 230 Ultrasonic humidifier, high-capacity system; 85 to 305 VAC
$2,934.99CAD / Each