This product is now discontinued, View Related Products
TriForest Culture Tubes, PC
Withstand multiple autoclave cycles
  • Polycarbonate (PC) is a safe and shatterproof alternative to glass
  • Keep samples secure—leakproof seals included
  • View samples through ultra-clear polycarbonate
These culture tubes are ideal for samples, fluids, and media that will ultimately be sterilized. Tubes feature a flat bottom and easily integrate into manual or automatic filling systems. Optically-clear polycarbonate is a safer alternative to glass with a wide range of chemical resistance.
Tubes are non-sterile and caps a packed separately.
Autoclavable
$830.99 - $1,068.21CAD / Case of 1500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Tube Capacity (mL) Tube Length (mm) Tube Outside Diameter (OD) (mm)
Availability Pricing
TriForest TPC13100 Culture Tubes, 13 mm x 100 mm, Flat Bottom, PC, Screw, packs of 250, 1500/CS RK-06019-45 Mfr # TPC13100
COMPARE
8 100 13
$830.99CAD / Case of 1500
TriForest TPC15103 Culture Tubes, 15 mm x 103 mm, Flat Bottom, PC, Screw, packs of 286, 1716/CS RK-06019-47 Mfr # TPC15103
COMPARE
11 103 15
$1,068.21CAD / Case of 1716
TriForest TPC16125 Culture Tubes, 16 mm x 125 mm, Flat Bottom, PC, Screw, packs of 200, 1200/CS RK-06019-48 Mfr # TPC16125
COMPARE
15 125 16
$1,025.48CAD / Case of 1200
TriForest TPC20125 Culture Tubes, 20 mm x 125 mm, Flat Bottom, PC, Screw, packs of 150, 900/CS RK-06019-49 Mfr # TPC20125
COMPARE
26 125 20
$839.83CAD / Case of 900