Trajan SGE Syringes
Designed for a vast variety of applications with outstanding performance and superior quality
Trajan SGE Syringes have consistently been the choice of global GC & LC manufacturers because of outstanding performance and superior quality in a vast variety of applications. The unique Plunger tip designed ensures a gas-tight seal providing the lowest syringe carryover and outstanding reproducibility.
Available range includes following variants:
GC Autosampler Syringes:
• SGE range of GC Autosampler syringes are designed and tested to meet critical autosampler specifications
• Wide range of fixed and removable needle options
• PTFE tipped replaceable plunge for gas and headspace samples
Gas-Tight Syringes:
• Constructed of borosilicate glass and interchangeable PTFE plunger tips
• Maximum inertness for liquid and gas samples
• Capacity varies from 10 µL to 10 mL
• High accuracy of dispensed volume
Jumbo Syringes:
• Designed for holding and dispensing large volumes of gas
• Heavy-duty acrylic barrels
• Easy access to sample for the addition of standards or removal of subsample via secondary port
• Plunger stem can be unscrewed for ease of transportation and storage
• Available in 500 mL, 1 L and 2 L volumes
$63.80 - $1,841.74CAD / Each
100 variations of this product are available.
See All Product Options Below
100 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Trajan SGE 000301 Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 23 G, Bevel Tip; 1/Ea 1599865 Mfr # 000301
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 23 G, Bevel Tip; 1/Ea
$184.54CAD / Each
Trajan SGE 000303 Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 26 G, Cone Tip; 1/Ea 1599866 Mfr # 000303
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 0.5 uL, 50 mm L, 26 G, Cone Tip; 1/Ea
$184.54CAD / Each
Trajan SGE 000350 Plunger-in-Needle Syringe with Repeating Adaptor, 0.5 uL, 23 G, 50 mm L, Cone Tip; 1/Ea 1599867 Mfr # 000350
COMPARE
Trajan SGE Plunger-in-Needle Syringe with Repeating Adaptor, 0.5 uL, 23 G, 50 mm L, Cone Tip; 1/Ea
$243.48CAD / Each
Trajan SGE 000400 Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 26 G; 1/Ea 1599872 Mfr # 000400
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 26 G; 1/Ea
$190.07CAD / Each
Trajan SGE 000415 Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 23-26 G; 1/Ea 1599874 Mfr # 000415
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 23-26 G; 1/Ea
$234.27CAD / Each
Trajan SGE 000440 Removable Needle Syringe, Compatible with Shimadzu Autosampler, 0.5uL, 42 mm L, 26 G; 1/Ea 1599875 Mfr # 000440
COMPARE
Trajan SGE Removable Needle Syringe, Compatible with Shimadzu Autosampler, 0.5uL, 42 mm L, 26 G; 1/Ea
$208.85CAD / Each
Trajan SGE 000445 Removable Needle Syringe, Compatible with Shimadzu Autosampler,  0.5 uL, 23 G; 1/Ea 1599876 Mfr # 000445
COMPARE
Trajan SGE Removable Needle Syringe, Compatible with Shimadzu Autosampler, 0.5 uL, 23 G; 1/Ea
$208.85CAD / Each
Trajan SGE 000475 Removable Needle CT Syringe, Compatible with PerkinElmer Autosampler, 0.5 uL, 26 G; 1/Ea 1599877 Mfr # 000475
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with PerkinElmer Autosampler, 0.5 uL, 26 G; 1/Ea
$239.06CAD / Each
Trajan SGE 000480 Removable Needle CT Syringe, Compatible with Thermo RSH Autosampler, 0.5 uL, 23 G; 1/Ea 1599879 Mfr # 000480
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with Thermo RSH Autosampler, 0.5 uL, 23 G; 1/Ea
$285.10CAD / Each
Trajan SGE 000490 Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 26 G; 1/Ea 1599880 Mfr # 000490
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 26 G; 1/Ea
$206.64CAD / Each
Trajan SGE 000492 Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 23 G; 1/Ea 1599881 Mfr # 000492
COMPARE
Trajan SGE Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 23 G; 1/Ea
$206.64CAD / Each
Trajan SGE 000500 Plunger-in-Needle Syringe, 1 uL, 50 mm L, 23 G, Cone Tip; 1/Ea 1599882 Mfr # 000500
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 1 uL, 50 mm L, 23 G, Cone Tip; 1/Ea
$155.81CAD / Each
Trajan SGE 000501 Plunger-in-Needle Syringe, Bevel Tip, 1 uL, 50 mm L, 23 G; 1/Ea 1599883 Mfr # 000501
COMPARE
Trajan SGE Plunger-in-Needle Syringe, Bevel Tip, 1 uL, 50 mm L, 23 G; 1/Ea
$156.92CAD / Each
Trajan SGE 000505 Plunger-in-Needle Syringe, 1 uL, 70 mm L, 23 G, Cone Tip; 1/Ea 1599884 Mfr # 000505
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 1 uL, 70 mm L, 23 G, Cone Tip; 1/Ea
$152.50CAD / Each
Trajan SGE 000506 Plunger-in-Needle Syringe, 1 uL, 70 mm L, 23 G, Bevel Tip; 1/Ea 1599885 Mfr # 000506
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 1 uL, 70 mm L, 23 G, Bevel Tip; 1/Ea
$152.50CAD / Each
Trajan SGE 000510 Plunger-in-Needle Syringe, 1 uL, 115 mm L, 23 G, Cone Tip; 1/Ea 1599886 Mfr # 000510
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 1 uL, 115 mm L, 23 G, Cone Tip; 1/Ea
$173.86CAD / Each
Trajan SGE 000553 Plunger-in-Needle Syringe with Repeating Adaptor, 1 uL, 70 mm L, 23 G, Cone Tip; 1/Ea 1599888 Mfr # 000553
COMPARE
Trajan SGE Plunger-in-Needle Syringe with Repeating Adaptor, 1 uL, 70 mm L, 23 G, Cone Tip; 1/Ea
$277.00CAD / Each
Trajan SGE 000570 Plunger-in-Needle Syringe, 1 uL, 115 mm L, 26 G, Cone Tip; 1/Ea 1599889 Mfr # 000570
COMPARE
Trajan SGE Plunger-in-Needle Syringe, 1 uL, 115 mm L, 26 G, Cone Tip; 1/Ea
$148.44CAD / Each
Trajan SGE 000680 Removable CT Needle Syringe, Compatible with Thermo RSH, 1 uL, 57 mm L, 23 G; 1/Ea 1599892 Mfr # 000680
COMPARE
Trajan SGE Removable CT Needle Syringe, Compatible with Thermo RSH, 1 uL, 57 mm L, 23 G; 1/Ea
$285.10CAD / Each
Trajan SGE 000790 Removable CT Needle Syringe, Compatible with Thermo RSH TriPlus and AS3000, 2 uL; 1/Ea 1599893 Mfr # 000790
COMPARE
Trajan SGE Removable CT Needle Syringe, Compatible with Thermo RSH TriPlus and AS3000, 2 uL; 1/Ea
$303.52CAD / Each
Showing 1 - 20 of 100