This product is now discontinued, View Related Products
Traceable® Thermohygrometers with Calibration
Monitor parameters with high and low alarm levels
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Audible alarm and flashing display actuate when high or low limits are exceeded
Thermohygrometers accurately measure humidity and temperature, and calculate dew point. Program high/low audible/visual alarms for temperature, humidity, and dew point. Choose from two models: universal or fast-response.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$663.03 - $901.71CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
COMPARE
9V battery (1) 95 140
$663.03CAD / Each
COMPARE
9V battery (1) 95 140
$901.71CAD / Each