Traceable® Standard Precalibrated Thermocouple Thermometers
Perfect for the lab to production floor
  • Includes A2LA accreditied NIST-Traceable Calibration Certificate
  • Reliable measurements, guaranteed
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of thermocouple probe errors
Handheld thermocouple thermometers provide accurate and reliable temperature measurement in an economic and compact design. Features include Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic off set function and auto power-off to save on battery power. Meters accept both type K or J thermocouple probes with a miniconnector.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$179.75 - $359.51CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
COMPARE
No 1 Type J, Type K
$179.75CAD / Each
COMPARE
Yes 2 Type J, Type K
$271.10CAD / Each
COMPARE
Yes 4 Type J, Type K
$359.51CAD / Each