Traceable® Remote-Monitoring Thermocouple Thermometers with Calibration
Ultra-thin cable lets you measure within doored equipment
  • Won’t affect door seal—ideal for use in remote measurement applications
  • For use in remote measurement applications
  • High-impact, chemical-resistant ABS case
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
This meter has a 10-ft. ribbon micro-cable that allows you to place the probe within an area and close the door without affecting the probe or the door seal. Accurately monitor temperature within freezers, water baths, heating blocks, incubators, refrigerators, and other areas. The fast-response 100 Ω platinum RTD probe can be used in liquids, air/gas, or frozen material.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$51.57CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Meter Measurement Type Types Of Probes Accepted Description
Availability Pricing
COMPARE
Thermocouple Type K Traceable® Remote-Monitoring Thermocouple Thermometer with Calibration; Fahrenheit
$51.57CAD / Each
COMPARE
Thermocouple Type K Traceable® Remote-Monitoring Thermocouple Thermometer with Calibration; Celsius
$51.57CAD / Each