Traceable® Metal Thermometers with Calibration
Waterproof metal construction—ideal for applications in wet labs, washdown areas, and outdoors
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Use these metal thermometers to record Max and Min temperatures in food and pharmaceutical labs, and cleanrooms. Metal housing and sealed construction provide IP67 rating for use in wet environments. Large, bright LCD with 3/8”H digits is easy to read. Features include selectable °F/°C units, Min/Max, Hold, and low-battery indicator.
Select from general-purpose thermometer with a 5”L stainless steel stem or dishwasher thermometer with a stand and hook. Dishwasher thermometer ensures proper cleaning is achieved by providing proof of achieving 180°F (82°C) during dishwashing.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$65.57 - $69.25CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
85 -20 Calibrated Metal Thermometer; General-Purpose
$65.57CAD / Each
COMPARE
85 -20 Calibrated Metal Thermometer; Dishwasher
$69.25CAD / Each