This product is now discontinued, View Related Products
Todd Systems Inc Step-down transformers
Power your instruments with 110/120 or 220/240 VAC outlets
  • Step-up models include NEMA 6-15P 230 VAC adapter for wiring convenience
  • Include rugged NEMA cord with three-prong plug
Select a step-down transformer to connect 110/120 VAC instruments to 220/240 VAC power sources; use a step-up transformer to connect 220/240 VAC instruments to 110/120 VAC power sources. All models run on 50/60 Hz.
$88.92 - $297.48CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Input (VAC) Output (VAC) Power Rating (watts)
Availability Pricing
Todd Systems SD-16 G Inc Step-down transformer, 100 Watts RK-01578-02 Mfr # SD-16 G
COMPARE
220240 110/120 100
$88.92CAD / Each
Todd Systems SD-12 G Inc Step-down transformer, 250 Watts RK-01578-05 Mfr # SD-12 G
COMPARE
220240 110/120 250
$123.40CAD / Each
Todd Systems SD-11 G Inc Step-down transformer, 500 Watts RK-01578-06 Mfr # SD-11 G
COMPARE
220240 110/120 500
$188.59CAD / Each
Todd Systems SD-13 G Inc Step-down transformer, 1000 Watts RK-01578-08 Mfr # SD-13 G
COMPARE
220240 110/120 1000
$240.75CAD / Each
Todd Systems SD-14 G Inc Step-down transformer, 1500 Watts RK-01578-09 Mfr # SD-14 G
COMPARE
220240 110/120 1500
$297.48CAD / Each