BRAND® Titrette® Bottletop Burettes (Burets)
Burette (burets) meets Class A standards for glass burettes!
  • Recirculation valve allows priming without wasting titrant
  • Mounts directly on bottle to reduce liquid transfer
Burette (burets) switches automatically between filling and titrating modes. View dispensed results on the LCD readout with a choice of resolution of 0.01µL, or 0.001µL for volumes below 20 mL. Unit can be easily and quickly disassembled for simple maintenance and cleaning, no servicing is required. Titrating tube can be adjusted horizontally and vertically. Suitable many common titrants with a concentration up to 1 mole/liter.
$2,011.18 - $2,202.72CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Volume (mL) Min Volume (mL)
Availability Pricing
BRAND Titrette®  Digital Bottletop Burette/Dispenser; 25 mL RK-07910-51 Mfr # 4760151
COMPARE
±0.2% 25 0.01
$2,202.72CAD / Each
BRAND Titrette®  Digital Bottletop Burette/Dispenser; 50 mL RK-07910-53 Mfr # 4760161
COMPARE
±0.2% 50 0.01
$2,202.72CAD / Each
BRAND Titrette®  Digital Bottletop Burette/Dispenser; 10mL RK-07910-55 Mfr # 4760141
COMPARE
±0.2% 10 0.01
$2,011.18CAD / Each