This product is now discontinued, View Related Products
Tight-Head Seamless Drums
No more heavy steel drums
  • Withstand rough handling—lightweight, rugged design
  • Easy access through 2" and 3⁄4" NPT bungs
  • Tough sides can support mixer
Tight-head drums meet HM-181 and IATA Specification Packaging Standards. These drums are constructed of durable LLDPE for a wide range of applications.
$162.51 - $309.71CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal) Wall thickness (in)
Availability Pricing
COMPARE
LLDPE 15 0.12
$162.51CAD / Each
COMPARE
LLDPE 30 0.188
$189.77CAD / Each
COMPARE
LLDPE 55 0.25
$309.71CAD / Each