This product is now discontinued, View Related Products
ThermoSafe Brands Infectious Substance Shippers 16538
Meet U.S. Department of Transportation (D.O.T.) and IATA requirements for transporting infectious substances
  • Pre-assembled, pretested, and qualified systems include everything you need to safely ship your infectious materials
  • Use infectious shipper overpack to protect frozen samples for more than four days
Infectious Substance Shippers allow you ship infectious materials safely. Foam containers are equipped with clear plastic press-lock bags and obsorbent in corrugated cartons. Shipper has a screw-cap aluminum can (6'' x 3'' dia), flexible foam insert with four pre-drilled holes for sterile tubes.
Shipper Overpack 03731-74 maintains frozen samples for more than four days. The overpack has a foam container and lid with cut out design.
$38.31 - $656.98CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Outside Height (in) Outside Length (in) Outside Width (in)
Availability Pricing
COMPARE
11 1/4 5 1/2 5 1/2
$95.03CAD / Each
COMPARE
14 3/4 14 1/4 14 1/4
$38.31CAD / Each
ThermoSafe INF2000 Infectious Substance Shipper, case of 6 RK-03731-76 Mfr # INF2000
COMPARE
6 3/4 4 4
$197.43CAD / Case of 6
ThermoSafe INF3000 Infectious Substance Shipper, case of 12 RK-03731-78 Mfr # INF3000
COMPARE
7 1/2 6 1/4 6 1/4
$656.98CAD / Case of 12