This product is now discontinued, View Related Products
Thermo Scientific Undercounter Refrigerators
  • Low-cost refrigerators designed for your laboratory applications
Refrigerators feature an easy-to-clean interior with epoxy-coated, steel-wire shelves that resist most acids, solvents, and chemicals. All refrigerators feature CFC-free insulation and coolant. Featuring adjustable hydraulic thermostat control and off-cycle defrost. Key lock protects the refrigerator from unauthorized entry.
$3,433.00 - $3,995.83CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature - Refrigerator (° F) Power (VAC)
Availability Pricing
Thermo Scientific 05LREETSA Undercounter Refrigerator, 5.6 cu ft, 120 VAC RK-44202-00 Mfr # 05LREETSA
COMPARE
53.6 115
$3,433.00CAD / Each
Thermo Scientific 05LREETSV Undercounter Refrigerator, 5.6 cu ft, 240 VAC RK-44202-05 Mfr # 05LREETSV
COMPARE
46.4 230
$3,995.83CAD / Each