This product is now discontinued, View Related Products
Thermo Scientific Steri-Cycle CO2 Incubators
Unique combination of HEPA air filtration and sterilization cycle provides complete contamination control
  • Large chamber provides ample room for samples
  • Stackable to preserve lab space
These reliable, direct-heat incubators feature an in-chamber HEPA air filtration system that continuously filters the entire chamber volume every 60 seconds and maintains cleanroom-like Class 100 air-quality conditions. Class 100 air quality conditions are achieved within five minutes following a routine door opening. Additionally, an on-demand 140°C high-temperature sterilization cycle eliminates contamination from all internal surfaces simplifying cleaning procedures.
Accurate microprocessor control of temperature and CO2 concentration is easy to program. Other programmable specifications include configurable audible on/off, an access code, a HEPA filter change reminder, remote alarm contacts, tracking low temperature and high and low CO2 alarms.
Select the TC sensor model when chamber temperature and relative humidity are relatively constant. Select the IR sensor model when temperature and relative humidity levels are changed frequently.
$12,700.62 - $25,393.88CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
COMPARE
6.5 115
$12,700.62CAD / Each
COMPARE
6.5 230
$25,393.88CAD / Each
COMPARE
6.5 115
$21,377.42CAD / Each
COMPARE
6.5 230
$14,574.77CAD / Each