This product is now discontinued, View Related Products
Thermo Scientific SnakeSkin Pleated Dialysis Tubes
Scale down your dialysis prep time for large sample volumes from 12 to 100 mL
  • The pleated format makes it easy to open and ready to use, streamlining dialysis preparation, save considerable time over dry, flat tubing.
  • Regenerated cellulose is ideal for desalting, buffer exchange and removal of small molecular weight contaminants.
The dialysis tubes are supplied in 8¡± sticks containing 35 feet of 22 mm internal diameter tubing. The amount of tubing required can be calculated using 3.7 mL sample per cm of dry tubing. To use it, a researcher simply pulls out the required length of tubing, cuts it off and applies a closure. The sample is then added through the other end of the dry tubing and the second closure is applied.
$253.42 - $481.06CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Thermo Scientific 68035 SnakeSkin Pleated Dialysis Tubing, 3500 MWCO, 22mm ID; 35ft RK-02904-00 Mfr # 68035
COMPARE
3500
$481.06CAD / Each
Thermo Scientific 68700 SnakeSkin Pleated Dialysis Tubing, 7000 MWCO, 22mm ID; 35ft RK-02904-05 Mfr # 68700
COMPARE
7000
$253.42CAD / Each
Thermo Scientific 68100 SnakeSkin Pleated Dialysis Tubing, 10000 MWCO, 22mm ID; 35ft RK-02904-10 Mfr # 68100
COMPARE
10000
$422.57CAD / Each