This product is now discontinued, View Related Products
Thermo Scientific™ Revco™ RDE Ultra-Low Temperature Freezers
Store up to 60,000 samples securely, silently and sustainably
  • H-drive Information Center (HIC) features easy-to-read temperature and simple capacitive touch buttons in a 5.6” display
  • Handle capacities between 300 to 600 2-inch storage boxes
  • Five inner doors allow easy sample access with limited external exposure to other contents
  • Security features include alarm settings, pass code, and padlock-compatible doors
  • Low peak variation ensures temperature protection throughout entire cabinet
  • Compatible with most racking systems and cryoboxes
With a small footprint, low noise output and lessened heat emissions that can reduce HVAC costs, these ultra-low temperature freezers provide a simple, sensible solution for your laboratory storage needs. No setup is needed; just plug the freezer in and it's ready to use.
Along with the HIC touch-screen interface, lab professionals will value single-handed door operation, the large eye-level display and shelf-level placement that doesn't require floor-level loading of racks. With SNAP-compliant natural hydrocarbon refrigerants, fast door-opening recovery times and estimated energy consumption as low as 10.7 kWh/day (depending on capacity), you'll also be making an environmentally-conscious choice.
$27,957.58 - $38,359.71CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Freezer Capacity (cu ft) Max Temperature - Freezer (° F) Min Temperature - Freezer (° F)
Availability Pricing
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 14.9 cu ft; 115 VAC 60 Hz RK-44146-00 Mfr # RDE30086FA
COMPARE
14.9 -58 -123
$27,957.58CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 14.9 cu ft; 208 to 230 VAC 60 Hz RK-44146-15 Mfr # RDE30086FD
COMPARE
14.9 -58 -123
$27,957.58CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 19.4 cu ft; 115 VAC 60 Hz RK-44146-16 Mfr # RDE40086FA
COMPARE
19.4 -58 -123
$30,558.11CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 19.4 cu ft; 208 to 230 VAC 60 Hz RK-44146-17 Mfr # RDE40086FD
COMPARE
19.4 -58 -123
$30,558.11CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 24.1 cu ft; 115 VAC 60 Hz RK-44146-18 Mfr # RDE50086FA
COMPARE
24.1 -58 -123
$33,917.44CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 24.1 cu ft; 208 to 230 VAC 60 Hz RK-44146-19 Mfr # RDE50086FD
COMPARE
24.1 -58 -123
$33,917.44CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 28.8 cu ft; 115 VAC 60 Hz RK-44146-40 Mfr # RDE60086FA
COMPARE
28.8 -58 -123
$38,359.71CAD / Each
Thermo Scientific Revco RDE Ultra-Low Temperature Freezer, 28.8 cu ft; 208 to 230 VAC 60 Hz RK-44146-41 Mfr # RDE60086FD
COMPARE
28.8 -58 -123
$38,359.71CAD / Each
Showing 1- 8 of 8