This product is now discontinued, View Related Products
Thermo Scientific™ Precision™ Refrigerated Incubator
Mmicroprocessor control ensure temperature accuracy and data integrity
  • Energy efficient — switch off refrigeration when set point is above ambient
  • Large capacity chamber supports 300 BOD bottles (300 mL) or over one hundred 4" (10-cm) pots or insect rearing cups
  • Temperature limit alarms, safety relay, and lockable door protect samples and organisms
  • CFC-free, foamed polyurethane insulation keeps temperature stable in changing or fluctuating ambient conditions
  • Access port allows passage of apparatus and probe cables
  • Chamber shelf height is adjustable to accommodate a variety of containers and apparatus
These refrigerated incubators feature microprocessor controls with easy-to-read display that shows actual temperature within 0.1 °C. Forced air circulation, delivers excellent temperature stability for applications requiring temperatures at or below ambient from -10 to 50 °C (14 to 122 °F).
Ideal for biochemical oxygen demand (BOD) testing, bacterial research, seed germination, fish/insect breeding and much more, having the largest usable capacity. Door includes six built-in shelves suitable for smaller sample bottles.
$10,741.01 - $14,501.10CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Thermo Scientific PR505755R Refrigerated Incubator, 20 cu ft, 120 VAC RK-39350-20 Mfr # PR505755R
COMPARE
20 50 120
$12,126.00CAD / Each
Thermo Scientific PR505750R Refrigerated Incubator, 20 cu ft, 230 VAC RK-39350-25 Mfr # PR505750R
COMPARE
20 50 230
$10,741.01CAD / Each
Thermo Scientific PR505755L Refrigerated Incubator with Day/Night Cycle, 17.8 cu ft, 120 VAC RK-39350-30 Mfr # PR505755L
COMPARE
17.8 50 120
$13,105.80CAD / Each
Thermo Scientific PR505750L Refrigerated Incubator with Day/Night Cycle, 17.8 cu ft, 230 VAC RK-39350-35 Mfr # PR505750L
COMPARE
17.8 50 230
$14,501.10CAD / Each