This product is now discontinued, View Related Products
Thermo Scientific Precision Compact Gravity Convection Ovens
Gentle drying with low turbulence
  • Doors open 180° for unhindered access
  • Circuit breaker protects the oven from power surges
  • Enamel-painted steel exterior and a stainless-steel interior are easy to clean
Compact Ovens are ideal for basic drying and baking applications with a temperature range of ambient +5 to 210°C. The small footprint conserves valuable bench top space or may be placed under laborotory counters. The pre-set high temperature safety thermostat ensures overtemperature protection, while pilot light visually indicates oven operation status. Ovens feature double wall aluminum interior chamber with 1'' silica-based insulation and powder coated, cold-rolled steel exterior.
$1,124.20 - $1,791.64CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific PR305215G Compact Gravity Oven, 0.7 Cu. Ft. (19.8 L), Ambient +5°C - 210°C, 120v RK-05014-01 Mfr # PR305215G
COMPARE
0.7 120
$1,124.20CAD / Each
Thermo Scientific PR305210G Compact Gravity Oven, 0.7 Cu. Ft. (19.8 L), Ambient +5°C - 210°C, 240v RK-05014-02 Mfr # PR305210G
COMPARE
0.7 240
$1,124.20CAD / Each
Thermo Scientific PR305225G Compact Gravity Oven; 1.7 cu ft, 120 V RK-05014-03 Mfr # PR305225G
COMPARE
1.7 120
$1,628.10CAD / Each
Thermo Scientific PR305220G Compact Gravity Oven; 1.7 cu ft, 240 V RK-05014-04 Mfr # PR305220G
COMPARE
1.7 240
$1,791.64CAD / Each