Thermo Scientific™ Orion™ ORP Standards
No hazardous chemicals or organics
  • Use as relative mV standard and calibrate offset of sample
  • Redox potential values are 215 mV for Ag/AgCl reference electrodes and 420 mV for standard hydrogen reference electrodes
These stable, ready-to-use standards require no mixing and contain no organics or hazardous chemicals — use straight out of the bottle. Choose from two convenient package sizes for use in the lab or field.
$173.12 - $250.11CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
Thermo Scientific 967901 ORP Standard, one 475 mL bottle RK-05730-55 Mfr # 967901
COMPARE
475 ORP Standard, one 475 mL bottle
$173.12CAD / Each
Thermo Scientific 967961 ORP Standard, 5 x 60 mL bottles RK-05730-57 Mfr # 967961
COMPARE
60 ORP Standard, 5 x 60 mL bottles
$250.11CAD / Each