Thermo Scientific™ Nunc™ Sterile Cell Culture Multidishes with Nunclon™ Delta Surface Treatment
Nunclon certified surface ensures optimum cell attachment and growth—use for scale up, cloning, viral titration, and fusion
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Flat well bottom and stackable
  • Raised well rims lower the risk of cross contamination
  • Alphanumerical codes for well identification
Round well styles feature vented lids for gas exchange and recessed areas around wells that act as a water reservoir to reduce evaporation from wells. Dishes with 6, 12, 24, and 48-well formats include lids with condensation rings to minimize evaporation. The 12 and 48-well dishes have a standard 96-well footprint. Rectangular well styles offer a larger culture area. They also accept standard glass cover slips: 24 x 60mm for 4-well plate; and 22 x 22mm for 8 well plate. Radiation sterilized.
$524.53 - $723.43CAD / Case of 75
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Culture Area (cm²) Max Working Well Volume (mL) Number of Wells
Availability Pricing
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 6-Well, 3 mL; 75/Cs RK-01930-19 Mfr # 140675
COMPARE
9.6 3 6
$524.53CAD / Case of 75
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 6-Well, 3 mL; 85/Cs RK-01930-21 Mfr # 140685
COMPARE
9.6 3 6
$568.73CAD / Case of 85
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 48-Well, 0.5 mL; 75/Cs RK-01930-24 Mfr # 150687
COMPARE
1.1 0.5 48
$723.43CAD / Case of 75
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 4-Well, 1 mL; 120/Cs RK-01930-25 Mfr # 176740
COMPARE
1.9 1 4
$708.70CAD / Case of 120
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 12-Well, 2 mL; 75/Cs RK-01930-29 Mfr # 150628
COMPARE
3.5 2 12
$548.10CAD / Case of 75
Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 24-Well, 1 mL; 85/Cs RK-01930-40 Mfr # 142485
COMPARE
1.9 1 24
$689.55CAD / Case of 85