This product is now discontinued, View Related Products
Thermo Scientific NEMA 4X and Explosion-Proof I/P (Current to Pressure) Transducers
Field selectable direct and reverse modes
  • Accuracies of ±0.15% and ±0.25% of span
  • Intrinsically safe input on all models
  • Explosion-proof enclosure (STD6000 models only)
  • Rugged NEMA 4X and IP65 enclosures on all models
Immune to shock and vibration, the Brandt current-to-pressure (I/P) transducer is built for rugged, industrial environments. The field-proven E-Pi transducer technology is a revolutionary breakthrough that provided the industry with its first “solid state” I/P transducer.
This advanced E-Pi technology uses a minimal amount of electrical energy and air consumption to convert a 4 to 20 input signal to a proportional pneumatic output signal. This pneumatic (backpressure) output is precisely modulated by a virtually weightless, low-mass membrane that is held in a continuously balanced position. The output of the E-Pi is fed into an integral volume booster to deliver a pneumatic output signal with an output capacity of 4.0 scfm. Overall performance, accuracy and repeatability are further enhanced via an internal feedback network that allows the transducer to quickly respond to input changes. These balanced supply and exhaust dynamics enhance stability while delivering accuracy of ±0.15% of span for superior process control.
$652.71 - $3,094.12CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Control Range (high) (PSI) Forward Flow Capacity (SCFM) Max Inlet Pressure (PSI)
Availability Pricing
COMPARE
15 4 20
$1,997.92CAD / Each
COMPARE
$652.71CAD / Each
COMPARE
30 4 35
$1,778.38CAD / Each
COMPARE
15 4 20
$3,094.12CAD / Each
COMPARE
$879.61CAD / Each
COMPARE
30 4 30
$2,103.26CAD / Each