Cole-Parmer® Essentials Narrow-Mouth Plastic Wash Bottles, FEP
Won't contaminate reagents
  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable
FEP bottles resist most chemicals and will not contaminate reagents—use to dispense strong solvents. A one-piece closure with spout prevents leaks.
$166.86 - $459.70CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 GL25 Fluorinated Ethylene Propylene (FEP)
$166.86CAD / Each
COMPARE
250 GL32 Fluorinated Ethylene Propylene (FEP)
$194.86CAD / Each
COMPARE
500 GL32 Fluorinated Ethylene Propylene (FEP)
$272.58CAD / Each
COMPARE
1000 GL32 Fluorinated Ethylene Propylene (FEP)
$459.70CAD / Each