This product is now discontinued, View Related Products

Thermo Scientific™ Nalgene™ Economy LDPE Wash Bottles

Eliminate pesky leaks
  • One-piece molded polypropylene closure and stem eliminate leaks
  • Easily cut stem to increase flow
These wash bottles feature a one-piece molded polypropylene (PP) closure and stem that eliminates leaks. The tip can be cut back to increase flow. Delivery tube is welded into the closure to prevent separation.
$55.99 - $79.56CAD / Pkg of 6

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2401-0125 Economy LDPE Wash Bottle, 125 mL, 6/pk RK-62304-20 Mfr # 2401-0125
COMPARE
125 24 mm Low Density Polyethyene (LDPE)
$55.99CAD / Pkg of 6
Thermo Scientific Nalgene 2401-0250 Economy LDPE Wash Bottle, 250 mL, 6/pk RK-62304-30 Mfr # 2401-0250
COMPARE
250 24 mm Low Density Polyethyene (LDPE)
$64.83CAD / Pkg of 6
Thermo Scientific Nalgene 2401-0500 Economy LDPE Wash Bottle, 500 mL, 6/pk RK-62304-40 Mfr # 2401-0500
COMPARE
500 28 mm Low Density Polyethyene (LDPE)
$79.56CAD / Pkg of 6
Thermo Scientific Nalgene 2401-1000 Economy LDPE Wash Bottle, 1 L, 4/pk RK-62304-50 Mfr # 2401-1000
COMPARE
1000 38-430 Low Density Polyethyene (LDPE)
$67.78CAD / Pkg of 4
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat