Thermo Scientific Nalgene Automatic Self-Zeroing Burettes (Burets)
No lubrication needed for PTFE stopcock
  • Won’t break— burette (buret) is made of clear acrylic body, PTFE stopcock, and PMP tip
Break-resistant burettes (burets) have clear acrylic body, polymethylpentene (PMP) tips and PTFE stopcocks. Burettes are individually calibrated to meet accuracy requirements for ASTM E 287. Unaffected by most dilute acids and bases (except HF and ammonium hydroxide). Use over full pH range. Not designed for use with alcohols or organic solvents.
Order support stand/rod and burette clamp separately.
$338.88 - $455.28CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL)
Availability Pricing
Thermo Scientific Nalgene 3640-0010 Self-Zeroing Burette, 10 mL; 1/Pk RK-06209-10 Mfr # 3640-0010
COMPARE
10 0.1
$338.88CAD / Each
Thermo Scientific Nalgene 3640-0025 Self-Zeroing Burette, 25 mL; 1/Pk RK-06209-20 Mfr # 3640-0025
COMPARE
25 0.1
$412.55CAD / Each
Thermo Scientific Nalgene 3640-0050 Self-Zeroing Burette, 50 mL; 1/Pk RK-06209-30 Mfr # 3640-0050
COMPARE
50 0.1
$455.28CAD / Each