This product is now discontinued, View Related Products
Thermo Scientific Multi-Blok Dry Heater, Blocks
Blocks measure 21-5/16"W x 2"H x 3-3/4"D. Solid anodized aluminum construction ensures maximum heat transfer between the heater base and tube contents. All blocks have 1-7⁄8" deep wells (except model 03130-08 with 1.1" deep wells and model 03137-00 with no wells). Blocks can accommodate a 1⁄4" dia thermometer.
$68.51 - $733.75CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mm) Description
Availability Pricing
Thermo Scientific 2070Q Heating Block for 6 mm Tubes, 30 Openings RK-03130-00 Mfr # 2070Q
COMPARE
30 6 Heating Block for 6 mm Tubes, 30 Openings
$108.29CAD / Each
Thermo Scientific 2068Q Heating Block for 8 mm Tubes, 30 Openings RK-03130-08 Mfr # 2068Q
COMPARE
30 8 Heating Block for 8 mm Tubes, 30 Openings
$146.90CAD / Each
Thermo Scientific 2069Q Heating Block for 10 mm Tubes, 20 Openings RK-03130-10 Mfr # 2069Q
COMPARE
20 10 Heating Block for 10 mm Tubes, 20 Openings
$73.67CAD / Each
Thermo Scientific 2071Q Heating Block for 10 mm Tubes, 24 Openings RK-03131-00 Mfr # 2071Q
COMPARE
24 10 Heating Block for 10 mm Tubes, 24 Openings
$72.93CAD / Each
Thermo Scientific 2066 Heating Block for 10 mm Tubes, 12 Openings RK-03131-10 Mfr # 2066
COMPARE
12 10 Heating Block for 10 mm Tubes, 12 Openings
$733.75CAD / Each
Thermo Scientific 2072Q Heating Block for 12 or 13 mm Tubes, 22 Openings RK-03132-00 Mfr # 2072Q
COMPARE
22 13 Heating Block for 12 or 13 mm Tubes, 22 Openings
$81.04CAD / Each
Thermo Scientific 2073Q Heating Block for 15 or 16 mm Tubes, 12 Openings RK-03133-00 Mfr # 2073Q
COMPARE
12 Heating Block for 15 or 16 mm Tubes, 12 Openings
$68.51CAD / Each
Thermo Scientific 2081Q Heating Block for 17 or 18 mm Tubes, 12 Openings RK-03133-18 Mfr # 2081Q
COMPARE
12 Heating Block for 17 or 18 mm Tubes, 12 Openings
$282.30CAD / Each
Thermo Scientific 2074Q Heating Block 20 mm Tubes, 8 Openings RK-03134-00 Mfr # 2074Q
COMPARE
8 20 Heating Block 20 mm Tubes, 8 Openings
$89.88CAD / Each
Thermo Scientific 2075Q Heating Block for 25 mm Tubes, 6 Openings RK-03135-00 Mfr # 2075Q
COMPARE
6 25 Heating Block for 25 mm Tubes, 6 Openings
$89.14CAD / Each
Thermo Scientific 2076Q Combination Heating Block, 3, 5, and 6 Openings RK-03136-00 Mfr # 2076Q
COMPARE
3 25 Combination Heating Block, 3, 5, and 6 Openings
$397.67CAD / Each
Thermo Scientific 2078Q Solid Heating Block for Dry Block Heaters RK-03137-00 Mfr # 2078Q
COMPARE
Solid Heating Block for Dry Block Heaters
$122.95CAD / Each
Showing 1- 12 of 12