This product is now discontinued, View Related Products
Thermo Scientific Multi-Blok Dry Block Heater, Microtiter Plate Blocks
Blocks accommodate any 6- to 96-well plate (round or flat bottom). Aluminum construction provides excellent heat transfer; model 03137-08 provides direct contact between the block heater and the bottom of microtiter plates for greater temperature uniformity. Color changes can easily be viewed against the white anodized surfaces. Finger indentations allow easy removal. Overall dimensions are 3-3/4"W x 1-3/4"H x 5-7/8"D.
$139.97 - $170.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Direct Contact Microtiter Heating Block RK-03137-08 Mfr # 2064Q
COMPARE
Direct Contact Microtiter Heating Block
$170.91CAD / Each
Thermo Scientific 2065Q Microtiter Plate Heating Block RK-03137-10 Mfr # 2065Q
COMPARE
Microtiter Plate Heating Block
$139.97CAD / Each