Thermo Scientific Low-Voltage Power Supplies
Highly compact, reliable and easy to use
Operating as a constant voltage power supply, very simple to use. The adjustment knob and hi–lo range switch control the output, which is variable in 10-volt increments from 20 to 250 volts. One, two-position switch toggles the bright LED display from indicating output voltage to output current. This model is equipped with an over current detector for maximum safety. With four-year warranty.
Model 28408-00 equipped with an over current detector for maximum safety. Model 28402-02 operates in constant voltage or constant current with a continously variable output. A dual function, auto ranging 24- hour up/down timer allows for more effective time management.
$1,326.05CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Height (in) Length (in) Width (in)
Availability Pricing
Thermo Scientific 105ECA-115 low-voltage power supply, 115V, 75W RK-28408-00 Mfr # 105ECA-115
COMPARE
4 1/2 6 8
$1,326.05CAD / Each