Thermo Scientific Locator Cryobiological Inventory Systems
Store samples up to 185 days without replenishing LN2
  • Compact size stores up to 3600 samples
  • Advanced vacuum insulation minimizes liquid nitrogen evaporation
Find the sample you need quickly and efficiently! All Thermo Scientific Locator® systems feature a double-walled, vacuum insulated vessel with a narrow-mouth neck that keeps liquid nitrogen losses to a minimum. Stainless steel racks fit snugly against the wall of the Thermo Scientific Locator® system to hold multiple racks. The rack handles rest in numbered slots for easy identification and retrieval. Each rack has individual storage locations for cryogenic boxes.
Use Thermo Scientific Locator® Jr. system (model 05094-60) to fit in tight spaces - this scaled-down version is only 26" in height! Thermo Scientific Locator® Jr. system holds four racks; each rack holds four 5-1/4" square cryogenic boxes. Select Locator 4 system (model 05094-40) to hold up to four racks; each rack holds nine 51⁄4" (13.3 cm) square cryogenic boxes. The Locator 8 system (model 05094-50) holds up to eight racks; each rack holds nine 3" (7.6 cm) square cryogenic boxes.
Order optional roller cart to easily transport your Locator system.
$11,566.11CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Static Holding Time (days) Tubes Held
Availability Pricing
Thermo Scientific CY50935 4 Cryobiological Inventory Systems RK-05094-40 Mfr # CY50935
COMPARE
112 3600
$11,566.11CAD / Each