Thermo Scientific Lindberg Blue M Split-Hinge Tube Furnaces
Maximum temperature of 1200°C
  • Ideal for annealing, crystal growing, calibration, and heat treating
  • Designed for vertical or horizontal operation
The split hinge design permits easy process tube installation, fast cooling, and overall ease of operation. These furnaces come equipped with energy-efficient Moldatherm® LGO™ heating elements that provide superior temperature uniformity, fast heat-up and cooldown and require little or no maintenance.
The heat-reflecting, element support assembly creates two highly effective insulating air spaces and the unique cabinet design helps keep the exterior surface temperature low. All models can be used with a variety of process tubes (sold separately) and come complete with one set of tube adapters.
All furnaces require an independent control module; sold separately.
$12,273.34CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Process Tube Length (in) Process Tube OD (in)
Availability Pricing
Thermo Scientific Lindberg HTF55347C Blue M Tube Furnace, 3 x 8" zones, 1"-3" OD; 208V RK-33903-13 Mfr # HTF55347C
COMPARE
208 / 240 24 1 to 3
$12,273.34CAD / Each