This product is now discontinued, View Related Products
Thermo Scientific™ I-Chem™ VOA Sample Vials
Certified and Processed vials meet or exceed EPA performance based standards for volatile organic analysis
  • Available as Certified to meet EPA Performance Based Specifications for Volatile Organic Analysis or Processed, which is the same as Certified vials but without certification documentation
These Volatile Organic Analysis (VOA) containers are pre-cleaned and certified to meet strict EPA requirements. Made of clear borosilicate glass, vials have a 0.125''-thick, I-CHEM blue, PTFE/silicone septa and open-top 24-414 polypropylene cap. Molded-in retainer ring holds septa securely in place without bonding to the cap.
$337.26 - $586.04CAD / Case of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Septum Type Tube Material Volume (mL)
Availability Pricing
I-Chem S226-0020 VOA Sample Vials, Processed, Glass, Polypropylene Cap with Unbonded Septa, 20 mL; 72/Cs RK-99536-10 Mfr # S226-0020
COMPARE
PTFE/silicone Borosilicate Glass 20
$362.45CAD / Case of 1
I-Chem S236-0040 VOA Sample Vials, Processed, Glass, Polypropylene Cap with Unbonded Septa, 40 mL; 72/Cs RK-99536-12 Mfr # S236-0040
COMPARE
PTFE/silicone Borosilicate Glass 40
$430.97CAD / Case of 1
I-Chem S236-0060 VOA Sample Vials, Processed, Glass, Polypropylene Cap with Unbonded Septa, 60 mL; 72/Cs RK-99536-16 Mfr # S236-0060
COMPARE
PTFE/silicone Borosilicate Glass 60
$492.48CAD / Case of 1
I-Chem S326-0020 VOA Sample Vials, Certified, Polypropylene Cap with Unbonded Septa, 20 mL; 72/Cs RK-99536-20 Mfr # S326-0020
COMPARE
PTFE/silicone Borosilicate Glass 20
$457.49CAD / Case of 72
I-Chem S336-0040 VOA Sample Vials, Certified, Glass, Polypropylene Cap with Unbonded Septa, 40 mL; 72/Cs RK-99536-22 Mfr # S336-0040
COMPARE
PTFE/silicone Borosilicate Glass 40
$538.89CAD / Case of 72
I-Chem S336-0060 VOA Sample Vials, Certified, Glass, Polypropylene Cap with Unbonded Septa, 60 mL; 72/Cs RK-99536-26 Mfr # S336-0060
COMPARE
PTFE/silicone Borosilicate Glass 60
$586.04CAD / Case of 72
I-Chem S336D0.5012 Certified Sample Vials, Field Ready, 40 mL; 72/Cs RK-99536-32 Mfr # PP140-40CEP.5HA
COMPARE
PTFE/silicone Borosilicate Glass 40
$337.26CAD / Case of 72
Showing 1- 7 of 7