This product is now discontinued, View Related Products

Thermo Scientific HyPURITY CN (Cyano) Drop-In Guard Cartridges

Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement—simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention. Sold in packs of 4.
$434.65 - $455.42CAD / Pkg of 4

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific HyPurity 22805-011001 CN Drop-In Guard Cartridges, 1mm ID, 5um; 4/Pk RK-01876-91 Mfr # 22805-011001
COMPARE
5
$434.65CAD / Pkg of 4
Thermo Scientific HyPurity 22805-012101 C18 Drop-In Guard Cartridge - Length 10mm, 2.1mm ID, 4pk RK-01876-92 Mfr # 22805-012101
COMPARE
$455.42CAD / Pkg of 4
Thermo Scientific HyPurity 22805-013001 C18 Drop-In Guard Cartridge - Length 10mm, 3.0mm ID RK-01876-93 Mfr # 22805-013001
COMPARE
$434.65CAD / Pkg of 4
Thermo Scientific HyPurity 22805-014001 C18 Drop-In Guard Cartridge - Length 10mm, 4.0mm / 4.6mm ID, 4pk RK-01876-94 Mfr # 22805-014001
COMPARE
$455.42CAD / Pkg of 4
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat