This product is now discontinued, View Related Products
Thermo Scientific HyPURITY ADVANCE Drop-In Guard Cartridges
Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement—simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention. Sold in packs of 4.
$425.66 - $504.49CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific HyPurity 21003-011001 Drop-In Guard Cartridges, 1.0mm ID, 3um; 4/Pk RK-01875-00 Mfr # 21003-011001
COMPARE
1 3
$425.66CAD / Each
Thermo Scientific HyPurity 21003-012101 Drop-In Guard Cartridges, 2.1mm ID, 3um; 4/Pk RK-01875-01 Mfr # 21003-012101
COMPARE
1 3
$504.49CAD / Each
Thermo Scientific HyPurity 21003-013001 Drop-In Guard Cartridges, 3.0mm ID, 3um; 4/Pk RK-01875-02 Mfr # 21003-013001
COMPARE
1 3
$425.66CAD / Each
Thermo Scientific HyPurity 21003-014001 Drop-In Guard Cartridges, 4.6/4.0mm ID, 3um; 4/Pk RK-01875-03 Mfr # 21003-014001
COMPARE
1 3
$504.49CAD / Each
Thermo Scientific HyPurity 21005-011001 Drop-In Guard Cartridges, 1.0mm ID, 5um; 4/Pk RK-01875-19 Mfr # 21005-011001
COMPARE
5
$455.42CAD / Pkg of 4
Thermo Scientific HyPurity 21005-012101 Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk RK-01875-20 Mfr # 21005-012101
COMPARE
5
$455.42CAD / Pkg of 4
Thermo Scientific HyPurity 21005-013001 Drop-In Guard Cartridges, 3.0mm ID, 5um; 4/Pk RK-01875-21 Mfr # 21005-013001
COMPARE
5
$455.42CAD / Pkg of 4
Thermo Scientific HyPurity 21005-014001 Drop-In Guard Cartridges, 4.6/4.0mm ID, 5um; 4/Pk RK-01875-22 Mfr # 21005-014001
COMPARE
5
$455.42CAD / Pkg of 4
Showing 1- 8 of 8