This product is now discontinued, View Related Products
Thermo Scientific™ Hypersil Green PAH Classical HPLC Columns
High-efficiency columns with proven results and reproducibility
Thermo Scientific Hypersil columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil Green PAH Columns are for the analysis of polyaromatic hydrocarbons. These columns provide complete resolution of the 16 PAHs according to EPA Method 610, including structurally similar pollutants. Stable in pH range of 2 to 9.
$823.26 - $1,831.42CAD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 31103-052130 Green PAH Column - Length 50mm, 2.1mm ID, 3µm particle size RK-01889-81 Mfr # 31103-052130
COMPARE
3
$873.72CAD / Each
Thermo Scientific Hypersil 31103-053030 Green PAH Column - Length 50mm, 3.0mm ID, 3µm particle size RK-01889-82 Mfr # 31103-053030
COMPARE
3
$873.72CAD / Each
Thermo Scientific Hypersil 31103-054030 Green PAH Column - Length 50mm, 4.0mm ID, 3µm particle size RK-01889-83 Mfr # 31103-054030
COMPARE
3
$829.15CAD / Each
Thermo Scientific Hypersil 31103-054630 Green PAH Column - Length 50mm, 4.6mm ID, 3µm particle size RK-01889-84 Mfr # 31103-054630
COMPARE
3
$829.15CAD / Each
Thermo Scientific Hypersil 31103-102130 Green PAH Column - Length 100mm, 2.1mm ID, 3µm particle size RK-01889-85 Mfr # 31103-102130
COMPARE
3
$951.07CAD / Each
Thermo Scientific Hypersil 31103-103030 Green PAH Column - Length 100mm, 3.0mm ID, 3µm particle size RK-01889-86 Mfr # 31103-103030
COMPARE
3
$951.07CAD / Each
Thermo Scientific Hypersil 31103-104030 Green PAH Column - Length 100mm, 4.0mm ID, 3µm particle size RK-01889-87 Mfr # 31103-104030
COMPARE
3
$903.56CAD / Each
Thermo Scientific Hypersil 31103-104630 Green PAH Column - Length 100mm, 4.6mm ID, 3µm particle size RK-01889-88 Mfr # 31103-104630
COMPARE
3
$903.56CAD / Each
Thermo Scientific Hypersil 31103-152130 Green PAH Column - Length 150mm, 2.1mm ID, 3µm particle size RK-01889-89 Mfr # 31103-152130
COMPARE
3
$1,004.48CAD / Each
Thermo Scientific Hypersil 31103-153030 Green PAH Column - Length 150mm, 3.0mm ID, 3µm particle size RK-01889-90 Mfr # 31103-153030
COMPARE
3
$1,004.48CAD / Each
Thermo Scientific Hypersil 31103-154030 Green PAH Column - Length 150mm, 4.0mm ID, 3µm particle size RK-01889-91 Mfr # 31103-154030
COMPARE
3
$954.02CAD / Each
Thermo Scientific Hypersil 31103-154630 Green PAH Column - Length 150mm, 4.6mm ID, 3µm particle size RK-01889-92 Mfr # 31103-154630
COMPARE
3
$954.02CAD / Each
Thermo Scientific Hypersil 31105-102130 Green PAH Column - Length 100mm, 2.1mm ID, 5µm particle size RK-01889-96 Mfr # 31105-102130
COMPARE
5
$861.93CAD / Each
Thermo Scientific Hypersil 31105-103030 Green PAH Column - Length 100mm, 3.0mm ID, 5µm particle size RK-01889-97 Mfr # 31105-103030
COMPARE
5
$861.93CAD / Each
Thermo Scientific Hypersil 31105-104030 Green PAH Column - Length 100mm, 4.0mm ID, 5µm particle size RK-01889-98 Mfr # 31105-104030
COMPARE
5
$823.26CAD / Each
Thermo Scientific Hypersil 31105-104630 Green PAH Column - Length 100mm, 4.6mm ID, 5µm particle size RK-01889-99 Mfr # 31105-104630
COMPARE
5
$823.26CAD / Each
Thermo Scientific Hypersil 31105-152130 Green PAH Column - Length 150mm, 2.1mm ID, 5µm particle size RK-01890-00 Mfr # 31105-152130
COMPARE
5
$910.92CAD / Each
Thermo Scientific Hypersil 31105-153030 Green PAH Column - Length 150mm, 3.0mm ID, 5µm particle size RK-01890-01 Mfr # 31105-153030
COMPARE
5
$910.92CAD / Each
Thermo Scientific Hypersil 31105-154030 Green PAH Column - Length 150mm, 4.0mm ID, 5µm particle size RK-01890-02 Mfr # 31105-154030
COMPARE
5
$872.25CAD / Each
Thermo Scientific Hypersil Green PAH Column - Length 150mm, 4.6mm ID, 5µm particle size RK-01890-03 Mfr # 31105-154630
COMPARE
5
$1,831.42CAD / Each
Showing 1 - 20 of 24