This product is now discontinued, View Related Products
Thermo Scientific Hypersil GOLD CN (Cyano) Columns
Thermo Scientific Hypersil GOLD CN Columns provide excellent peak shape and selectivity in reversed and normal phase separations, with lower hydrophobicity when compared to alkyl chain phases. These columns extend the range of available selectivities, and can be used in the analysis of compounds including steroids. Stable in a pH range of 2 to 8.
$734.48 - $1,206.71CAD / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 25802-021030 GOLD CN Column - Length 20mm, 1.0mm ID, 1.9µm particle size RK-01883-78 Mfr # 25802-021030
COMPARE
1.9
$937.81CAD / Each
Thermo Scientific Hypersil 25802-022130 GOLD CN Column - Length 20mm, 2.1mm ID, 1.9µm particle size RK-01883-79 Mfr # 25802-022130
COMPARE
1.9
$937.81CAD / Each
Thermo Scientific Hypersil 25802-023030 GOLD CN Column - Length 20mm, 3.0mm ID, 1.9µm particle size RK-01883-80 Mfr # 25802-023030
COMPARE
1.9
$937.81CAD / Each
Thermo Scientific Hypersil 25802-031030 GOLD CN Column - Length 30mm, 1.0mm ID, 1.9µm particle size RK-01883-81 Mfr # 25802-031030
COMPARE
1.9
$990.85CAD / Each
Thermo Scientific Hypersil 25802-032130 GOLD CN Column - Length 30mm, 2.1mm ID, 1.9µm particle size RK-01883-82 Mfr # 25802-032130
COMPARE
1.9
$990.85CAD / Each
Thermo Scientific Hypersil 25802-033030 GOLD CN Column - Length 30mm, 3.0mm ID, 1.9µm particle size RK-01883-83 Mfr # 25802-033030
COMPARE
1.9
$990.85CAD / Each
Thermo Scientific Hypersil 25802-051030 GOLD CN Column - Length 50mm, 1.0mm ID, 1.9µm particle size RK-01883-84 Mfr # 25802-051030
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-052130 GOLD CN Column - Length 50mm, 2.1mm ID, 1.9µm particle size RK-01883-85 Mfr # 25802-052130
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-053030 GOLD CN Column - Length 50mm, 3.0mm ID, 1.9µm particle size RK-01883-86 Mfr # 25802-053030
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-101030 GOLD CN Column - Length 100mm, 1.0mm ID, 1.9µm particle size RK-01883-87 Mfr # 25802-101030
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-102130 GOLD CN Column - Length 100mm, 2.1mm ID, 1.9µm particle size RK-01883-88 Mfr # 25802-102130
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-103030 GOLD CN Column - Length 100mm, 3.0mm ID, 1.9µm particle size RK-01883-89 Mfr # 25802-103030
COMPARE
1.9
$1,042.42CAD / Each
Thermo Scientific Hypersil 25802-152130 GOLD CN Column - Length 150mm, 2.1mm ID, 1.9µm particle size RK-01883-90 Mfr # 25802-152130
COMPARE
1.9
$1,206.71CAD / Each
Thermo Scientific Hypersil 25802-202130 GOLD CN Column - Length 200mm, 2.1mm ID, 1.9µm particle size RK-01883-91 Mfr # 25802-202130
COMPARE
1.9
$1,192.34CAD / Each
Thermo Scientific Hypersil 25803-031030 GOLD CN Column - Length 30mm, 1.0mm ID, 3µm particle size RK-01883-96 Mfr # 25803-031030
COMPARE
3
$926.03CAD / Each
Thermo Scientific Hypersil 25803-032130 GOLD CN Column - Length 30mm, 2.1mm ID, 3µm particle size RK-01883-97 Mfr # 25803-032130
COMPARE
3
$847.94CAD / Each
Thermo Scientific Hypersil 25803-033030 GOLD CN Column - Length 30mm, 3.0mm ID, 3µm particle size RK-01883-98 Mfr # 25803-033030
COMPARE
3
$847.94CAD / Each
Thermo Scientific Hypersil 25803-034030 GOLD CN Column - Length 30mm, 4.0mm ID, 3µm particle size RK-01883-99 Mfr # 25803-034030
COMPARE
3
$805.94CAD / Each
Thermo Scientific Hypersil 25803-034630 GOLD CN Column - Length 30mm, 4.6mm ID, 3µm particle size RK-01884-00 Mfr # 25803-034630
COMPARE
3
$805.94CAD / Each
Thermo Scientific Hypersil 25803-051030 GOLD CN Column - Length 50mm, 1.0mm ID, 3µm particle size RK-01884-01 Mfr # 25803-051030
COMPARE
3
$926.03CAD / Each
Showing 1 - 20 of 50